missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99845-100ul
This item is not returnable.
View return policy
Description
FLAP Polyclonal antibody specifically detects FLAP in Human samples. It is validated for ELISA
Specifications
| FLAP | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein | |
| Produced in rabbits immunized with purified, recombinant Human FLAP (Accession#: NP_001620.2; Met1-Pro161) | |
| 100 μg | |
| Immunology, Inflammation | |
| 241 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction