Learn More
Macherey-Nagel™ Flash Chromatography Kit II
Product Code. 10566531 Shop All Macherey-Nagel Products
Click to view available options
Type:
Flash Chromatography Kit II
Unit Size:
Each
Product Code. 10566531
Brand: Macherey-Nagel™ 727451
Specifications
Type Flash Chromatography Kit II
Includes Includes One 450 x 40 mm I.D. Glass Column, One 1-L eluent Reservoir, 100 g Silica 60 (40 to 63 μm), Sea Sand, Silanized Glass Fiber Wadding, 1 m PTFE Tubing
For Use With (Application) Low Pressure Flash Chromatography