missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLASH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA130057
This item is not returnable.
View return policy
Description
PA1-30057 detects FLASH from human samples.
This protein is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| FLASH | |
| Polyclonal | |
| Unconjugated | |
| CASP8AP2 | |
| AA387232; CASP8 associated protein 2; CASP8AP2; CASP8-associated protein 2; caspase 8 associated protein 2; CED-4; D4Ertd659e; FLASH; FLASH homolog RIP25; FLICE associated huge; FLICE-associated huge protein; FLJ11208; human FLASH; KIAA1315; mKIAA1315; RIP25; RP11-63K6.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UKL3 | |
| CASP8AP2 | |
| N-terminus of the FLASH protein. Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction