Learn More
Invitrogen™ FLASH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590692
Description
Immunogen sequence: TVGAFIDLTQ DASSEAKSEG NHPALAVEDL GCGVIQVDED NCKEEKAQVA NRPLKCIVEE TYIDLTTESP SSCEVKKDEL KSEPGSNCDN SELPGTLHNS HKKRRNISDL NHPHKKQRKE TDLTNKEKTK KPTQDSCENT EAHQKKASKK KAPPVTKDPS SLKATPGIKD SSAALATSTS LSAKNVIKKK GEIIILWTRN DDREILLECQ KRGPSFKTFA YLAAKLDKNP NQVSERFQQL MKLFEKSKCR; Positive Samples: U-87MG, K-562; Cellular Location: Cytoplasm, Mitochondrion, Nucleus, PML body.
This protein is highly similar to FLASH, a mouse apoptotic protein identified by its interaction with the death-effector domain (DED) of caspase 8. Studies of FLASH protein suggested that this protein may be a component of the death-inducing signaling complex that includes Fas receptor, Fas-binding adapter FADD, and caspase 8, and plays a regulatory role in Fas-mediated apoptosis. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| FLASH | |
| Polyclonal | |
| Unconjugated | |
| CASP8AP2 | |
| AA387232; CASP8 associated protein 2; CASP8AP2; CASP8-associated protein 2; caspase 8 associated protein 2; CED-4; D4Ertd659e; FLASH; FLASH homolog RIP25; FLICE associated huge; FLICE-associated huge protein; FLJ11208; human FLASH; KIAA1315; mKIAA1315; RIP25; RP11-63K6.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9994 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.85 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UKL3 | |
| CASP8AP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1733-1982 of human CASP8AP2 (NP_0362471). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.