Learn More
BUCHI FlashPure Chromatography Solid Loader
Product Code. 16350316 Shop All Buchi Products
Change view
Click to view available options
Size (Metric) Tube Rack:
20 g
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Tube Rack unitSize
16350316 20 g Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16350316 Supplier BUCHI Supplier No. 148634340
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Type Empty Solid Loader
Includes Frits
Size (Metric) Tube Rack 20 g
Max. Pressure 10 to 14 bar (145 to 200 psi)