Learn More
BUCHI FlashPure Chromatography Solid Loader
Product Code. p-9455008 Shop All Buchi Products
Click to view available options
Size (Metric) Tube Rack:
20 g
Unit Size:
Pack of 20
Product Code. 16350316
Brand: BUCHI 148634340
Only null left
Add to basket
Add to basket
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polypropylene
Type Empty Solid Loader
Includes Frits
Size (Metric) Tube Rack 20 g
Max. Pressure 10 to 14 bar (145 to 200 psi)