missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimble Chase™ Flat Bottom Borosilicate Glass Serum Vials
Description
- Borosilicate glass
- Flat bottom
- 100/Pack
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| For Use With (Application) | Ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples |
| Material | Borosilicate Glass |
| No. per Pack | 100Pack |
| Packaging | 100Pack |
| Quantity | 100Pack |
| Height (Metric) | 65 mm |
| Diameter (Metric) | 30 mm |
| Shape | Flat Bottom |
| Product Type | Serum Vial |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?