Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Two Angled Side Necks, and One Vertical Side Neck
€ 200.34 - € 725.04
Specifications
Material | Borosilicate Glass, Duran Glass |
---|---|
For Use With (Application) | Reaction vessels |
Product Type | Flange lid |
Product Code | Brand | Includes | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Closure Size | Price | Quantity & Availability | ||||
10861841
|
Lenz Laborglasinstrumente™
06104241 |
Laboratory Flange 100 | Center Neck (NS): 29/32 |
€ 252.28
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
10169529
|
Lenz Laborglasinstrumente™
06104264 |
Laboratory Flange 100 | Center Neck (NS): 45/40 |
€ 293.62
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10053473
|
Lenz Laborglasinstrumente™
06104444 |
Laboratory Flange 150 | Center Neck (NS): 29/32 |
€ 395.38
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
10179529
|
Lenz Laborglasinstrumente™
06104464 |
Laboratory Flange 150 | Center Neck (NS): 45/40 |
€ 420.82
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
11552364
|
Lenz Laborglasinstrumente™
06104564 |
Laboratory Flange 200 | Center Neck (NS): 45/40 |
€ 725.04
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
11542364
|
Lenz Laborglasinstrumente™
06104544 |
Laboratory Flange 200 | Center Neck (NS): 29/32 |
€ 708.08
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
11532364
|
Lenz Laborglasinstrumente™
06104111 |
Laboratory Flange 60 | Center Neck (NS): 14/23 |
€ 200.34
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
10101984
|
Lenz Laborglasinstrumente™
06104141 |
Laboratory Flange 60 | Center Neck (NS): 29/32 |
€ 207.76
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
Specifications
Borosilicate Glass, Duran Glass | |
Reaction vessels | |
Flange lid |