Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck and Two Angled Side Necks
Product Code. 10192454
Click to view available options
Height (Metric):
75 mm
85 mm
95 mm
110 mm
125 mm
150 mm
165 mm
Includes:
Laboratory Flange 100
Laboratory Flange 140
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
Product Code. 10192454
Brand: Lenz Laborglasinstrumente™ 06103141
Only null left
Add to basket
Add to basket
Spécification
Material Borosilicate Glass, Duran Glass
For Use With (Application) Reaction vessels
Height (Metric) 85 mm
Includes Laboratory Flange 60
Product Type Flange lid