missing translation for 'onlineSavingsMsg'
Learn More

Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)), Alexa Fluor™ 700 - Chimeric, Novus Biologicals™

Product Code. 18688749 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18688749 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18688749 Supplier Novus Biologicals Supplier No. NBP252666AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flavivirus group antigen
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone D1-4G2-4-15 (4G2)
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias Envelope Protein, Protein E
Host Species Rabbit
Immunogen This recombinant Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) - Chimeric was prepared from Dengue Virus type 2 antigens.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.