missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)), DyLight 350 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52666UV
This item is not returnable.
View return policy
Description
Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Neutralization, CyTOF
Specifications
| Flavivirus group antigen | |
| Monoclonal | |
| DyLight 350 | |
| Envelope Protein, Protein E | |
| This recombinant Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) - Chimeric was prepared from Dengue Virus type 2 antigens. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| D1-4G2-4-15 (4G2) | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction