missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52709PECY55
This item is not returnable.
View return policy
Description
Flavivirus group antigen Monoclonal antibody specifically detects Flavivirus group antigen in Virus samples. It is validated for Flow Cytometry
Specifications
| Flavivirus group antigen | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| Envelope Protein, Protein E | |
| This recombinant Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) was prepared from Dengue Virus type 2 antigens. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Flow Cytometry | |
| D1-4G2-4-15 (4G2) | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction