missing translation for 'onlineSavingsMsg'
Learn More

Flavopiridol Hydrochloride, MedChemExpress

Product Code. 16452768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452768 10 mg 10mg
16445507 5 mg 5mg
16429466 50 mg 50mg
16434226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452768 Supplier Medchem Express Supplier No. HY10006
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flavopiridol Hydrochloride (Alvocidib Hydrochloride) is a broad inhibitor of CDK, competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Flavopiridol Hydrochloride (Alvocidib Hydrochloride) is a broad inhibitor of CDK, competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Specifications

Chemical Name or Material Flavopiridol Hydrochloride
CAS 131740-09-5
Color pale Brown
Purity Grade Notes Research
Molecular Formula C21H21Cl2NO5
Quantity 10 mg
Synonym Alvocidib HydrochlorideL86-8275 HydrochlorideHMR-1275 Hydrochloride
Solubility Information H2O : ≥ 20 mg/mL (45.63 mM) ∣DMF : 7.69 mg/mL (17.55 mM; Need ultrasonic)
SMILES O=C1C2=C(C=C(C([C@]3([H])[C@H](O)CN(C)CC3)=C2OC(C4=CC=CC=C4Cl)=C1)O)O.[H]Cl
Molecular Weight (g/mol) 438.3
Formula Weight 438.3
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.