missing translation for 'onlineSavingsMsg'
Learn More

Flavopiridol Hydrochloride, MedChemExpress

Product Code. p-200095815
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452768

Brand: Medchem Express HY10006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flavopiridol Hydrochloride (Alvocidib Hydrochloride) is a broad inhibitor of CDK, competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Flavopiridol Hydrochloride (Alvocidib Hydrochloride) is a broad inhibitor of CDK, competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Specifications

Chemical Name or Material Flavopiridol Hydrochloride
CAS 131740-09-5
Color pale Brown
Purity Grade Notes Research
Molecular Formula C21H21Cl2NO5
Quantity 10 mg
Synonym Alvocidib HydrochlorideL86-8275 HydrochlorideHMR-1275 Hydrochloride
Solubility Information H2O : ≥ 20 mg/mL (45.63 mM) ∣DMF : 7.69 mg/mL (17.55 mM; Need ultrasonic)
SMILES O=C1C2=C(C=C(C([C@]3([H])[C@H](O)CN(C)CC3)=C2OC(C4=CC=CC=C4Cl)=C1)O)O.[H]Cl
Molecular Weight (g/mol) 438.3
Formula Weight 438.3
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.