missing translation for 'onlineSavingsMsg'
Learn More

Flecainide acetate, MedChemExpress

Product Code. 16447338
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16447338

Brand: Medchem Express HY17429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flecainide acetate (R-818) is a class 1C antiarrhythmic drug especially used for the management of supraventricular arrhythmia; works by blocking the Nav1.5 sodium channel in the heart, causing prolongation of the cardiac action potential.

Flecainide acetate (R-818) is a class 1C antiarrhythmic drug especially used for the management of supraventricular arrhythmia

Specifications

Chemical Name or Material Flecainide acetate
CAS 54143-56-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H24F6N2O5
Quantity 10 mg
Synonym R-818
Solubility Information DMSO : 50 mg/mL (105.40 mM; Need ultrasonic) ∣H2O : 20 mg/mL (42.16 mM; Need ultrasonic)
SMILES O=C(NCC1NCCCC1)C2=CC(OCC(F)(F)F)=CC=C2OCC(F)(F)F.CC(O)=O
Molecular Weight (g/mol) 474.39
Formula Weight 474.39
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.