Learn More
Eppendorf™ Flex Bucket Adapters for IsoRack
Add flexibility to your liquid handling applications with these IsoRack adapters
€ 170.00 - € 178.00
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | Flex bucket |
No. of Places | 96 |
No. per Pack | 2 Pack |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10224032
|
Eppendorf™
5825708008 |
0.5 mL |
€ 170.00
Pack of 2 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10171362
|
Eppendorf™
5825709004 |
1.5 to 2 mL |
€ 178.00
Pack of 2 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- IsoRack adapters to support 0.5 mL and 1.5/2.0 mL IsoRacks during centrifugation
- Adapters can be used in Eppendorf plate rotors A-2-DWP and A-4-81- MTP/Flex
Specifications
Adapter | |
Flex bucket | |
96 | |
2 Pack |