Learn More
OHAUS™ Flex System Clamp
Shop All Ohaus Products
Click to view available options
Length (Metric) Arm:
305 mm
457 mm
Includes:
Base Plate
Lab Frame Clamp
Shelf Clamp
Unit Size:
Each
Description
- Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
- Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifications
Specifications
| Type | Flex System Clamp |
| Material | Nickel-plated |
| Includes | Shelf Clamp |
| Description | Clamp, Specialty, Flex, CLS-FLEXBL |
| Diameter (Metric) | 13 mm |
| Length (English) Arm | 18 in. |
| Length (Metric) Arm | 457 mm |
| Size (Metric) Grip | 0 to 43 mm |
| Jaw Type | 3-Prong Dual Adjust |