Learn More
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For U1060
Flexible temperature sensor for positioning in chamber or in load. Memmert™ PT100 Temperature Sensor is designed with a 4-pin connector for external temperature recording.
Brand:  Memmert™ U1060H8
Additional Details : Weight : 0.01100kg
Product Code. 16316087
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: 4-Pin Connector for Temperature Recording
Description
Description
  • Can be positioned in chamber or in load
  • Includes 4-pin connector, according to NAMUR NE 28, for external temperature recording (load temperature) max. 3 sensors
  • For use in Memmert™ universal ovens
Specifications
Specifications
Secondary/Additional PT 100 Sensor (up to 3 can be attached in total)
Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108
Temperature Sensor
Load/Chamber Temperature Measurement