Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF450plus
Product Code. 17579405 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF450plus Oven
Unit Size:
Each
Product Code. 17579405
Brand: Memmert™ UF450plusH8
Only null left
Add to basket
Add to basket
Flexible temperature sensor for positioning in chamber or in load.
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
For Use With (Equipment) UF450plus Oven
Type Temperature Sensor