missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Antibody (FLI1/4371R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21020AF700
This item is not returnable.
View return policy
Description
FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FLI1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| Recombinant full-length human FLI1 protein | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 2313 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FLI1/4371R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction