Learn More
Flibanserin N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Flibanserin N-Oxide |
| CAS | 2652583-35-0 |
| Molecular Formula | C20 H21 F3 N4 O2 |
| Quantity | 250 mg |
| Synonym | Flibanserin N-Oxide, 1-(2-(2-Oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)ethyl)-4-(3-(trifluoromethyl)phenyl)piperazine 1-Oxide |
| InChI Formula | InChI=1S/C20H21F3N4O2/c21-20(22,23)15-4-3-5-16(14-15)25-8-11-27(29,12-9-25)13-10-26-18-7-2-1-6-17(18)24-19(26)28/h1-7,14H,8-13H2,(H,24,28) |
| SMILES | [O-][N+]1(CCN2C(=O)Nc3ccccc23)CCN(CC1)c4cccc(c4)C(F)(F)F |
| IUPAC Name | 3-[2-[1-oxido-4-[3-(trifluoromethyl)phenyl]piperazin-1-ium-1-yl]ethyl]-1H-benzimidazol-2-one |
| Molecular Weight (g/mol) | 406.401 |
| Formula Weight | 406.162 |