missing translation for 'onlineSavingsMsg'
Learn More

Flibanserin N-Oxide, TRC

Product Code. 30406312
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406312 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406312 Supplier Toronto Research Chemicals Supplier No. TRCF40127525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flibanserin N-Oxide
CAS 2652583-35-0
Molecular Formula C20 H21 F3 N4 O2
Quantity 25 mg
Synonym Flibanserin N-Oxide, 1-(2-(2-Oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)ethyl)-4-(3-(trifluoromethyl)phenyl)piperazine 1-Oxide
InChI Formula InChI=1S/C20H21F3N4O2/c21-20(22,23)15-4-3-5-16(14-15)25-8-11-27(29,12-9-25)13-10-26-18-7-2-1-6-17(18)24-19(26)28/h1-7,14H,8-13H2,(H,24,28)
SMILES [O-][N+]1(CCN2C(=O)Nc3ccccc23)CCN(CC1)c4cccc(c4)C(F)(F)F
IUPAC Name 3-[2-[1-oxido-4-[3-(trifluoromethyl)phenyl]piperazin-1-ium-1-yl]ethyl]-1H-benzimidazol-2-one
Molecular Weight (g/mol) 406.401
Formula Weight 406.162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.