missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ11506 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18422DL594
This item is not returnable.
View return policy
Description
FLJ11506 Polyclonal antibody specifically detects FLJ11506 in Human, Mouse samples. It is validated for Western Blot
Specifications
| FLJ11506 | |
| Polyclonal | |
| Western Blot | |
| AAGAB, alpha- and gamma-adaptin-binding protein p34, KPPP1, p34, PPKP1A | |
| Recognizes region between residues Between Between 265 and 315.. | |
| 0.1 mL | |
| Signal Transduction | |
| 79719 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction