missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ21963 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98578UV
This item is not returnable.
View return policy
Description
FLJ21963 Polyclonal antibody specifically detects FLJ21963 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FLJ21963 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| acyl-CoA synthetase short-chain family member 3, acyl-CoA synthetase short-chain family member 3, mitochondrial, EC 6.2.1, EC 6.2.1.1, FLJ21963 | |
| Produced in rabbits immunized with E. coli-derived Human FLJ21963 fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 79611 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction