missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ35220 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00284131-B01P
This item is not returnable.
View return policy
Description
FLJ35220 Polyclonal antibody specifically detects FLJ35220 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| FLJ35220 | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:2000 | |
| AAH64545.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 284131 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| EC 3.1, hypothetical protein FLJ35220, hypothetical protein LOC284131, MGC74455 | |
| FLJ35220 (AAH64545.1, 1 a.a. - 264 a.a.) full-length human protein. MALEAAGGPPEETLSLWKREQARLKAHVVDRDTEAWQRDPAFSGLQRVGGVDVSFVKGDSVRACASLVVLSFPELEVLLVDGNGVLHHRGFGVACHLGVLTDLPCVGVAKKLLQVDGLENNALHKEKIRLLQTRGDSFPLLGDSGTVLGMALRSHDRSTRPLYISVGHRMSLEAAVRLTCCCCRFRIPEPVRQADICSREHIRKSLGLPGPPTPRSPKAQRPVACPKGDSGESSGEGQPPQDHSPGPRTAPRPGSQEQAGKDWQ | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction