Learn More
FLJ40504 Mouse anti-Human, Clone: OTI5A6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies CF810103
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FLJ40504 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene.Spécification
| FLJ40504 | |
| Monoclonal | |
| Unconjugated | |
| KRT18P55 | |
| KRT18P55 | |
| Full length human recombit protein of human FLJ40504 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5A6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| keratin 18 pseudogene 55 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 284085 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.