Learn More
Scotch-Brite™ Floor Pads, Green
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 25
Pack of 5
Specifications
Specifications
| Product Type | Floor Pad |
| Color | Green |
| Diameter (Metric) | 480 mm |
| For Use With (Application) | Scrubbing |
| For Use With (Equipment) | Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines |
| Style | Round |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion