Learn More
Invitrogen™ Flotillin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581935
Description
Immunogen sequence: HQRAIMAHMT VEEIYKDRQK FSEQVFKVAS SDLVNMGISV VSYTLKDIHD DQDYLHSLGK ARTAQVQKDA RIGEAEAKRD AGIREAKAKQ EKVSAQYLSE IEMAKAQRDY ELKKAAYDIE VNTRRAQADL AYQLQVAKTK QQIEEQR Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Specifications
| Flotillin 1 | |
| Polyclonal | |
| Unconjugated | |
| FLOT1 | |
| DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10211, 14251, 64665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O08917, O75955, Q9Z1E1 | |
| FLOT1 | |
| Recombinant protein corresponding to Human FLOT1. Recombinant protein control fragment (Product #RP-94763). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.