missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flotillin 1 Polyclonal Antibody

Description
For Western Blot, this antibody has non-specific bands at 35 kDa. Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with many species based on sequence homology. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Specifications
Specifications
| Antigen | Flotillin 1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | FLOT1 |
| Gene Accession No. | O08917, O75955 |
| Gene Alias | DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 |
| Gene Symbols | FLOT1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?