missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLRT1 Antibody (017), DyLight 594 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89894DL594
This item is not returnable.
View return policy
Description
FLRT1 Monoclonal antibody specifically detects FLRT1 in Human samples. It is validated for ELISA
Specifications
| FLRT1 | |
| Monoclonal | |
| DyLight 594 | |
| fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FLRT1 (Q9NZU1-1; Met1-Pro524). | |
| 0.1 mL | |
| Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 23769 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 017 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction