missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLRT2 Antibody (11B5G9), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06631FR
This item is not returnable.
View return policy
Description
FLRT2 Monoclonal antibody specifically detects FLRT2 in Human samples. It is validated for ELISA
Specifications
| FLRT2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 11B5G9 | |
| ELISA | |
| Fibronectin Leucine Rich Transmembrane Protein 2, Fibronectin-Like Domain-Containing Leucine-Rich Transmembrane Protein 2, KIAA0405, Leucine-Rich Repeat Transmembrane Protein FLRT2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FLRT2 (O43155; Met 1-Ser 539). | |
| 0.1 mL | |
| Cell Biology, Signal Transduction, Stem Cells | |
| 23768 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction