missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLRT3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36566JF646
This item is not returnable.
View return policy
Description
FLRT3 Polyclonal antibody specifically detects FLRT3 in Human samples. It is validated for Western Blot
Specifications
| FLRT3 | |
| Polyclonal | |
| Western Blot | |
| fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3 | |
| The immunogen recognized this antibody maps to a region between residue 375 to 425 of human Fibronectin Leucine Rich Transmembrane Protein 3 using the numbering given in entry NP_938205.1. | |
| 0.1 mL | |
| Signal Transduction | |
| 23767 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction