missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (113308) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7681JF585
This item is not returnable.
View return policy
Description
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| Flt-3/Flk-2/CD135 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Flt-3/Flk-2, Asn28-Ser544 (predicted), Accession # Q00342 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 113308 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cell Cycle and Replication, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase | |
| 2322 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction