missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (12B6), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-94155AF647
This item is not returnable.
View return policy
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Flt-3/Flk-2/CD135 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FLT3 | |
| Mouse Flt-3/Flk-2/CD135 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| This recognizes mouse Flt3/CD135 Antibody (12B6) in western blot. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 12B6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| Rat | |
| Protein G purified | |
| Cell Cycle and Replication, Phospho Specific | |
| 2322 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction