missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (MM0294-9G23), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12311R
This item is not returnable.
View return policy
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot.
Specifications
| Flt-3/Flk-2/CD135 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FLT3 | |
| Human recombinant Flt-3/Flk-2/CD135 | |
| 0.1 mL | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
| Western Blot, Flow Cytometry | |
| MM0294-9G23 | |
| Western Blot, Flow Cytometry | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication, Phospho Specific | |
| 2322 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction