missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3 Ligand/FLT3L Antibody (008), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89408UV
This item is not returnable.
View return policy
Description
Flt-3 Ligand/FLT3L Monoclonal antibody specifically detects Flt-3 Ligand/FLT3L in Human samples. It is validated for ELISA
Specifications
| Flt-3 Ligand/FLT3L | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 008 | |
| ELISA | |
| FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Flt-3 Ligand/FLT3L (P49771-1; Thr27-Pro185). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 2323 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction