missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT1 Monoclonal Antibody (OTI11G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806786S
This item is not returnable.
View return policy
Description
FLT1 Monoclonal antibody specifically detects FLT1 in Human samples. It is validated for Western BlotSpecifications
| FLT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10451 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6696 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI11G5 | |
| Unconjugated | |
| SPP1 | |
| AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1 | |
| Human recombinant protein fragment corresponding to amino acids 17-314 of human SPP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction