missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT1 Monoclonal Antibody (OTI11G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806786
This item is not returnable.
View return policy
Description
Receptors for VEGF, VEGFB and PGF have a tyrosine-protein kinase activity. The VEGF-kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability. VEGF and its high-affinity binding receptors, the tyrosine kinases FLK1 and FLT1, are thought to be important for the development of embryonic vasculature. It has been shown that an alternately spliced form of FLT1 produces a soluble protein, termed sFLT1, which binds vascular endothelial growth factor with high affinity, playing an inhibitory role in angiogenesis.Specifications
| FLT1 | |
| Monoclonal | |
| Unconjugated | |
| FLT1 | |
| FLT, FLT-1, VEGFR-1, VEGFR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2321 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI11G5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17948 | |
| FLT1 | |
| Human recombit protein fragment corresponding to amino acids 781-1338 of human FLT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction