Learn More
FLT4 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806894S
Encompass_Preferred
 View more versions of this product
Product Code. 17577193
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
FLT4 Monoclonal antibody specifically detects FLT4 in Human samples. It is validated for Western Blot
Specifications
Specifications
FLT4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P09917
Mouse
Affinity chromatography
RUO
240
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G7
Unconjugated
ALOX5
AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3
Human recombinant protein fragment corresponding to amino acids 18-364 of human ALOX5 produced in E.coli.