Learn More
Fluazifop-butyl D9 100 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Fluazifop-butyl D9 (n-butyl D9) |
| CAS | 2714409-29-5 |
| Solvent or Matrix | Acetone |
| Molecular Formula | C19 2H9 H11 F3 N O4 |
| Quantity | 1 mL |
| Synonym | Fluazifop-butyl D9 |
| InChI Formula | InChI=1S/C19H20F3NO4/c1-3-4-11-25-18(24)13(2)26-15-6-8-16(9-7-15)27-17-10-5-14(12-23-17)19(20,21)22/h5-10,12-13H,3-4,11H2,1-2H3/i1D3,3D2,4D2,11D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OC(=O)C(C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1 |
| IUPAC Name | 1,1,2,2,3,3,4,4,4-nonadeuteriobutyl 2-[4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate |
| Molecular Weight (g/mol) | 392.42 |
| Show More |