Learn More
Fluensulfone, MedChemExpress
Product Code. 16486857
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Product Code. 16486857
Brand: Medchem Express HY107771
MedChemExpress Fluensulfone is a new nematicide for chemical control of plant parasitic nematodes.
Fluensulfone is a new nematicide for chemical control of plant parasitic nematodes.
Specifications
Chemical Name or Material Fluensulfone
Physical Form Solid
CAS 318290-98-1
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C7H5ClF3NO2S2
For Use With (Application) Cancer-programmed cell death
Synonym MCW-2
Solubility Information DMSO : 100 mg/mL (342.82 mM; Need ultrasonic) ∣H2O : 0.73 mg/mL (2.50 mM; Need ultrasonic)
SMILES F/C(F)=C(F)\CCS(=O)(C1=NC=C(Cl)S1)=O
Molecular Weight (g/mol) 291.7
Formula Weight 291.7
Percent Purity 98.75%
Grade Research
Show More Show Less
Research purposes only