missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunixin Meglumine EP Impurity D, Mikromol™
Description
Flunixin Meglumine EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-[(2-Methyl-3-(trifluoromethyl)phenyl)amino]pyridine-3-carboxylate (Flunixin Ethyl Ester) |
| CAS | 54396-42-8 |
| Analyte or Component Names | Ethyl 2-[(2-Methyl-3-(trifluoromethyl)phenyl)amino]pyridine-3-carboxylate (Flunixin Ethyl Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 F3 N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl 2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]pyridine-3-carboxylate (Flunixin Ethyl Ester), Flunixin Meglumine Imp. D (EP), 2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid ethyl ester, Flunixin Meglumine Impurity D, Flunixin Impurity D |
| InChI Formula | InChI=1S/C16H15F3N2O2/c1-3-23-15(22)11-6-5-9-20-14(11)21-13-8-4-7-12(10(13)2)16(17,18)19/h4-9H,3H2,1-2H3,(H,20,21) |
| SMILES | CCOC(=O)c1cccnc1Nc2cccc(c2C)C(F)(F)F |
| IUPAC Name | ethyl 2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?