missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fluorescein

Product Code. 16419895
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419895 Supplier MedChemExpress Supplier No. HYD0251
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fluorescein (Uranine) is widely used as a fluorescent tracer in medicinal and biological applications and tumor infected tissues tracer.

Fluorescein (Uranine) is a representative green fluorophore that has been widely used as a scaffold of practically useful green fluorescent probes.

Specifications

Quantity 250 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 332.31 g/mol
Solubility DMSO : 250 mg/mL (752.31 mM; Need ultrasonic) ∣ H2O : 1 mg/mL (3.01 mM; ultrasonic and warming and heat to 80°C)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 2321-07-5
Synonym Uranine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.