missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Antibody (4-4-20 (enhanced)), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52691AF488
This item is not returnable.
View return policy
Description
Fluorescein Monoclonal antibody specifically detects Fluorescein in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF
Specifications
| Fluorescein | |
| Monoclonal | |
| Alexa Fluor 488 | |
| FITC, fluorescein isothyocianate | |
| Fluorescein I-bovine serum albumin (FlI-BSA). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Control, CyTOF | |
| 4-4-20 (enhanced) | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction