missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-2082
This item is not returnable.
View return policy
Description
Fluorescein Polyclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Fluorescein | |
| Polyclonal | |
| Biotin | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Fluorescein | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:2000-1:10000, ELISA 1:10000-1:50000, Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| FITC, fluorescein isothyocianate | |
| Fluorescein conjugated to Goat IgG | |
| 0.2 mg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction