missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Antibody, Rhodamine - F(ab')2, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97384
This item is not returnable.
View return policy
Description
Fluorescein Polyclonal antibody specifically detects Fluorescein in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
| Fluorescein | |
| Polyclonal | |
| Rhodamine | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Reconstitute with 1.0 mL deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot >1:10000, Immunocytochemistry/ Immunofluorescence 1:500-1:2500, Fluorophore-linked immunosorbent assay >1:20000 | |
| FITC, fluorescein isothyocianate | |
| Fluorescein conjugated to Goat IgG | |
| 1 mg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction