missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Fluoresceinamine, isomer II, 95%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Melting Point | 285°C |
| CAS | 51649-83-3 |
| Molecular Formula | C20H13NO5 |
| MDL Number | MFCD00005051 |
| Beilstein | 51708 |
| Synonym | 6-Aminofluorescein |
| InChI Key | YOAWSYSKQHLFPM-UHFFFAOYSA-N |
| SMILES | C1=CC2=C(C=C1N)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O |
| IUPAC Name | 5-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
| Molecular Weight (g/mol) | 347.326 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction