Learn More
Fluorescent Brightener ER-II, 1-(2-Cyanostyryl)-4-(4-cyanostyryl)benzene, TRC
Specifications
Specifications
| Chemical Name or Material | Fluorescent Brightener ER-II, 1-(2-Cyanostyryl)-4-(4-cyanostyryl)benzene |
| CAS | 13001-38-2 |
| Molecular Formula | C24H16N2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C24H16N2/c25-17-22-13-11-20(12-14-22)6-5-19-7-9-21(10-8-19)15-16-23-3-1-2-4-24(23)18-26/h1-16H/b6-5+,16-15+ |
| SMILES | N#Cc1ccc(\C=C\c2ccc(\C=C\c3ccccc3C#N)cc2)cc1 |
| Molecular Weight (g/mol) | 332.41 |
| Formula Weight | 332.1313 |