missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals

Catalog Number p-4258862
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
164298-25-3
C9H16FN2·F6P
316.2
MNJUGQKOHJQOCK-UHFFFAOYSA-N
btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
10935980
1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
164298-25-3
C9H16FN2·F6P
316.2
MNJUGQKOHJQOCK-UHFFFAOYSA-N
btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
10935980
1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Melting Point 152.0°C to 154.0°C
Color White
Elemental Analysis 32.82 to 35.55% C, 8.51 to 9.21% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 316.2
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'