Learn More
Fluquinconazole, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Fluquinconazole |
| CAS | 136426-54-5 |
| CAS Max % | ≤ 100% |
| Density | 1.58 g/cm3 (20°C) |
| Color | White |
| Melting Point | 191°C to 193°C |
| Synonym | 3-(2,4-Dichlorophenyl)-6-fluoro-2-(1H-1,2,4-trizol-1-yl)quinazolin-4(3H)-one |
| Molecular Weight (g/mol) | 376.17 g/mol |
| Quantity | 100 mg |
| Grade | Fungicides and Bactericides |